1-(4-Chlorobenzenesulphonyl)-2-methyl-4,5-dihydro-1H-imidazole, tech

Lieferant: Apollo Scientific

OR33391-1G
APOSOR33391-1GEA 1570 EUR
APOSOR33391-1G
1-(4-Chlorobenzenesulphonyl)-2-methyl-4,5-dihydro-1H-imidazole, tech
1-(4-Chlorobenzenesulphonyl)-2-methyl-4,5-dihydro-1H-imidazole
Formel: C₁₀H₁₁ClN₂O₂S
Molecular Weight: 258,725 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00980765
CAS-Nummer: 181481-30-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR