Methyl 4-(2-methylbenzoyl)-1H-pyrrole-2-carboxylate, tech

Lieferant: Apollo Scientific

OR33392-1G
APOSOR33392-1GEA 1570 EUR
APOSOR33392-1G
Methyl 4-(2-methylbenzoyl)-1H-pyrrole-2-carboxylate, tech
Methyl 4-(2-methylbenzoyl)-1H-pyrrole-2-carboxylate
Formel: C₁₄H₁₃NO₃
Molecular Weight: 243,258 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571494
CAS-Nummer: 478249-31-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR