Methyl 4-(2-phenylacetyl)-1H-pyrrole-2-carboxylate, tech
Lieferant: Apollo Scientific
OR33394-1G
APOSOR33394-1GEA
1570
EUR
APOSOR33394-1G
Methyl 4-(2-phenylacetyl)-1H-pyrrole-2-carboxylate, tech
Methyl 4-(2-phenylacetyl)-1H-pyrrole-2-carboxylate
Formel:
C₁₄H₁₃NO₃ Molecular Weight: 243,258 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02571496 CAS-Nummer: 478249-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...