2-(2-Propynyloxy)benzenecarbaldehyde oxime 95%

Lieferant: Apollo Scientific

OR33397-1G OR33397-5G
APOSOR33397-1GEA 0 EUR
APOSOR33397-1G APOSOR33397-5G
2-(2-Propynyloxy)benzenecarbaldehyde oxime 95%
2-(2-Propynyloxy)benzenecarbaldehyde oxime
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03001307
CAS-Nummer: 55241-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR