3-Amino-5-cyanobenzeneboronic acid hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Amino-5-cyanophenyl)boronic acid hydrochloride
OR3340-250MG
APOSOR3340-250MGEA
46.4
EUR
APOSOR3340-250MG
3-Amino-5-cyanobenzeneboronic acid hydrochloride 97%
3-Amino-5-cyanobenzeneboronic acid hydrochloride
Formel:
C₇H₇BN₂O₂·ClH Molecular Weight: 198,415 g/mol Schmelzpunkt: 123…125 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD08436033 CAS-Nummer: 913835-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...