2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzonitrile, tech

Lieferant: Apollo Scientific

OR33403-1G
APOSOR33403-1GEA 1570 EUR
APOSOR33403-1G
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzonitrile, tech
2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzonitrile
Formel: C₁₃H₁₂N₂
Molecular Weight: 196,248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01964837
CAS-Nummer: 124678-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR