N-(4-Methoxyphenyl)-4,6-dimethylpyrimidin-2-amine, tech
Lieferant: Apollo Scientific
OR33405-1G
APOSOR33405-1GEA
1570
EUR
APOSOR33405-1G
N-(4-Methoxyphenyl)-4,6-dimethylpyrimidin-2-amine, tech
N-(4-Methoxyphenyl)-4,6-dimethylpyrimidin-2-amine
Formel:
C₁₃H₁₅N₃O Molecular Weight: 229,278 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00439512 CAS-Nummer: 23951-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...