4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)pentanoic acid, tech

Lieferant: Apollo Scientific

OR33406-1G
APOSOR33406-1GEA 1570 EUR
APOSOR33406-1G
4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)pentanoic acid, tech
4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)pentanoic acid
Formel: C₁₄H₁₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03305892
CAS-Nummer: 866144-51-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR