3-(Cyclohex-3-en-1-yl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech

Lieferant: Apollo Scientific

OR33407-1G
APOSOR33407-1GEA 1570 EUR
APOSOR33407-1G
3-(Cyclohex-3-en-1-yl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid, tech
3-(Cyclohex-3-en-1-yl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Formel: C₁₇H₁₉NO₃
Molecular Weight: 285,338 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305894
CAS-Nummer: 866144-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR