4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)hexanoic acid, tech
Lieferant: Apollo Scientific
OR33408-1G
APOSOR33408-1GEA
1570
EUR
APOSOR33408-1G
4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)hexanoic acid, tech
4-Methyl-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)hexanoic acid
Formel:
C₁₅H₁₉NO₃ Molecular Weight: 261,316 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03305896 CAS-Nummer: 866144-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...