4-Bromophenyl methyl sulphoxide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-(methylsulphinyl)benzene

OR3341-250MG
APOSOR3341-250MGEA 21.9 EUR
APOSOR3341-250MG
4-Bromophenyl methyl sulphoxide 98%
4-Bromophenyl methyl sulphoxide
Formel: C₇H₇BrOS
Molecular Weight: 219,099 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00159074
CAS-Nummer: 934-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR