Methyl-3-formyl-1H-indole-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-formyl-1H-indole-2-carboxylate

OR33422-1G
APOSOR33422-1GEA 72.5 EUR
APOSOR33422-1G
Methyl-3-formyl-1H-indole-2-carboxylate 95%
Methyl-3-formyl-1H-indole-2-carboxylate
Formel: C₁₁H₉NO₃
Molecular Weight: 203.2 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03848053
CAS-Nummer: 18450-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR