5,5-Dimethyl-2-phenyl-1,2,4-triazolidine-3-thione, tech

Lieferant: Apollo Scientific

OR33424-1G
APOSOR33424-1GEA 1570 EUR
APOSOR33424-1G
5,5-Dimethyl-2-phenyl-1,2,4-triazolidine-3-thione, tech
5,5-Dimethyl-2-phenyl-1,2,4-triazolidine-3-thione
Formel: C₁₀H₁₃N₃S
Molecular Weight: 207,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00684882
CAS-Nummer: 39263-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR