2-[4-(4-Methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Lieferant: Apollo Scientific
OR33425-1G
APOSOR33425-1GEA
1570
EUR
APOSOR33425-1G
2-[4-(4-Methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[4-(4-Methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel:
C₁₁H₁₀N₄O Molecular Weight: 214,223 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04126410 CAS-Nummer: 866149-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...