2-[4-(4-Bromophenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
Lieferant: Apollo Scientific
OR33426-1G
APOSOR33426-1GEA
1570
EUR
APOSOR33426-1G
2-[4-(4-Bromophenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile, tech
2-[4-(4-Bromophenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetonitrile
Formel:
C₁₀H₇BrN₄O Molecular Weight: 279,093 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04126417 CAS-Nummer: 866149-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...