2-(5-Oxo-4-phenyl-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetonitrile, tech

Lieferant: Apollo Scientific

OR33427-1G
APOSOR33427-1GEA 1570 EUR
APOSOR33427-1G
2-(5-Oxo-4-phenyl-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetonitrile, tech
2-(5-Oxo-4-phenyl-4,5-dihydro-1H-1,2,4-triazol-1-yl)acetonitrile
Formel: C₁₀H₈N₄O
Molecular Weight: 200,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04126418
CAS-Nummer: 866149-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR