(2E)-3-(Dimethylamino)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33432-1G
APOSOR33432-1GEA 1570 EUR
APOSOR33432-1G
(2E)-3-(Dimethylamino)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one
Formel: C₁₁H₁₅NOS
Molecular Weight: 209,308 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05668971
CAS-Nummer: 67382-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR