3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol, tech
Lieferant: Apollo Scientific
OR33434-1G
APOSOR33434-1GEA
1570
EUR
APOSOR33434-1G
3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol, tech
3,5-Dimethyl-2-[(morpholin-4-yl)methyl]phenol
Formel:
C₁₃H₁₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05668993 CAS-Nummer: 14944-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...