2,4-Dimethyl-6-[(morpholin-4-yl)methyl]phenol hydrochloride, tech

Lieferant: Apollo Scientific

OR33436-1G
APOSOR33436-1GEA 1570 EUR
APOSOR33436-1G
2,4-Dimethyl-6-[(morpholin-4-yl)methyl]phenol hydrochloride, tech
2,4-Dimethyl-6-[(morpholin-4-yl)methyl]phenol hydrochloride
Formel: C₁₃H₁₉NO₂·ClH
Molecular Weight: 257,756 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05974977
CAS-Nummer: 1052543-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR