Methyl-2-chlor-5-iodnicotinat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-chloro-5-iodonicotinate, Methyl 2-chloro-5-iodopyridine-3-carboxylate

OR33437-5G
APOSOR33437-5GEA 148 EUR
APOSOR33437-5G
Methyl-2-chlor-5-iodnicotinat 95%
Methyl-2-chlor-5-iodnicotinat
Formel: C₇H₅ClINO₂
Molecular Weight: 297.48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659607
CAS-Nummer: 78686-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR