6-(4-Formylphenyl)nicotinaldehyde 95%

Lieferant: Apollo Scientific

OR33438-1G
APOSOR33438-1GEA 0 EUR
APOSOR33438-1G
6-(4-Formylphenyl)nicotinaldehyde 95%
6-(4-Formylphenyl)nicotinaldehyde
Formel: C₁₃H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06659608
CAS-Nummer: 885950-11-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR