Methyl-4-(5-formyl-2-pyridinyl)benzenecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-(5-formylpyridin-2-yl)benzoate

OR33439-1G
APOSOR33439-1GEA 671 EUR
APOSOR33439-1G
Methyl-4-(5-formyl-2-pyridinyl)benzenecarboxylate 95%
Methyl-4-(5-formyl-2-pyridinyl)benzenecarboxylate
Formel: C₁₄H₁₁NO₃
Molecular Weight: 241,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659609
CAS-Nummer: 834884-66-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR