6-(3-Thienyl)nicotinaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 6-(Thiophen-3-yl)pyridine-3-carbaldehyde

OR33440-1G
APOSOR33440-1GEA 671 EUR
APOSOR33440-1G
6-(3-Thienyl)nicotinaldehyde 95%
6-(3-Thienyl)nicotinaldehyde
Formel: C₁₀H₇NOS
Molecular Weight: 189,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115422
CAS-Nummer: 834884-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR