2-(3-Thienyl)nicotinaldehyde 97%

Lieferant: Apollo Scientific

Synonyme: 2-(Thiophen-3-yl)pyridine-3-carbaldehyde

OR33441-1G
APOSOR33441-1GEA 671 EUR
APOSOR33441-1G
2-(3-Thienyl)nicotinaldehyde 97%
2-(3-Thienyl)nicotinaldehyde
Formel: C₁₀H₇NOS
Molecular Weight: 189,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659610
CAS-Nummer: 885950-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR