N-{3-[3-(1-Pyrrolidinyl)-1-propynyl]phenyl}acetamide 95%

Lieferant: Apollo Scientific

OR33443-1G
APOSOR33443-1GEA 538 EUR
APOSOR33443-1G
N-{3-[3-(1-Pyrrolidinyl)-1-propynyl]phenyl}acetamide 95%
N-{3-[3-(1-Pyrrolidinyl)-1-propynyl]phenyl}acetamide
Formel: C₁₅H₁₈N₂O
Molecular Weight: 242,32 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795473
CAS-Nummer: 885950-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR