tert-Butyl-4-hydroxydihydro-2(3H)-isoxazolecarboxylate 95%
Lieferant: Apollo Scientific
OR33444-1G
APOSOR33444-1GEA
894
EUR
APOSOR33444-1G
tert-Butyl-4-hydroxydihydro-2(3H)-isoxazolecarboxylate 95%
tert-Butyl-4-hydroxydihydro-2(3H)-isoxazolecarboxylate
Formel:
C₈H₁₅NO₄ Molecular Weight: 189,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07698596 CAS-Nummer: 262444-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...