Ethyl 5-formyl-1-benzofuran-2-carboxylate, tech

Lieferant: Apollo Scientific

OR33447-1G
APOSOR33447-1GEA 1570 EUR
APOSOR33447-1G
Ethyl 5-formyl-1-benzofuran-2-carboxylate, tech
Ethyl 5-formyl-1-benzofuran-2-carboxylate
Formel: C₁₂H₁₀O₄
Molecular Weight: 218.205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11501717
CAS-Nummer: 10035-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR