3-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%
Lieferant: Apollo Scientific
Synonyme:
(E)-N-{[3-(1H-Pyrazol-1-yl)phenyl]methylidene}hydroxylamine
OR33459-1G
APOSOR33459-1GEA
336
EUR
APOSOR33459-1G
3-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%
3-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime
Formel:
C₁₀H₉N₃O Molecular Weight: 187.2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09152715 CAS-Nummer: 1017782-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...