4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbonitrile 95%

Lieferant: Apollo Scientific

Synonyme: 4-[(5-Bromopyrimidin-2-yl)oxy]benzonitrile

OR33460-1G
APOSOR33460-1GEA 671 EUR
APOSOR33460-1G
4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbonitrile 95%
4-[(5-Bromo-2-pyrimidinyl)oxy]benzenecarbonitrile
Formel: C₁₁H₆BrN₃O
Molecular Weight: 276,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03646339
CAS-Nummer: 887430-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR