2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%

Lieferant: Apollo Scientific

Synonyme: (E)-N-{[2-(1H-Pyrazol-1-yl)phenyl]methylidene}hydroxylamine

OR33462-1G
APOSOR33462-1GEA 336 EUR
APOSOR33462-1G
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime 95%
2-(1H-Pyrazol-1-yl)benzenecarbaldehyde oxime
Formel: C₁₀H₉N₃O
Molecular Weight: 187.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09152718
CAS-Nummer: 1017782-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR