[5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-yl]-N-methylmethanamine 95%

Lieferant: Apollo Scientific

OR33471-1G
APOSOR33471-1GEA 894 EUR
APOSOR33471-1G
[5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-yl]-N-methylmethanamine 95%
[5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-yl]-N-methylmethanamine
Formel: C₁₀H₁₀ClN₃O
Molecular Weight: 223,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09972216
CAS-Nummer: 1017782-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR