2'-(Methylsulphinyl)[1,1'-biphenyl]-4-carbaldehyde 95%

Lieferant: Apollo Scientific

OR33473-1G OR33473-5G
APOSOR33473-1GEA 336 EUR
APOSOR33473-1G APOSOR33473-5G
2'-(Methylsulphinyl)[1,1'-biphenyl]-4-carbaldehyde 95%
2'-(Methylsulphinyl)[1,1'-biphenyl]-4-carbaldehyde
Formel: C₁₄H₁₂O₂S
Molecular Weight: 244.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09972222
CAS-Nummer: 1171155-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR