1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone 95%
Lieferant: Apollo Scientific
OR33474-1G
APOSOR33474-1GEA
538
EUR
APOSOR33474-1G
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone 95%
1-(3-Bromo-1-benzofuran-2-yl)-1-ethanone
Formel:
C₁₀H₇BrO₂ Molecular Weight: 239,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09972223 CAS-Nummer: 99661-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...