N-(2-Bromo-5-methoxy-4-methylphenyl)-2-chloroacetamide 95%

Lieferant: Apollo Scientific

OR33480-1G
APOSOR33480-1GEA 538 EUR
APOSOR33480-1G
N-(2-Bromo-5-methoxy-4-methylphenyl)-2-chloroacetamide 95%
N-(2-Bromo-5-methoxy-4-methylphenyl)-2-chloroacetamide
Formel: C₁₀H₁₁BrClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11840961
CAS-Nummer: 1135282-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR