Methyl-4-propionyl-1H-pyrrole-2-carboxylate 97%
Lieferant: Apollo Scientific
OR33482-1G
APOSOR33482-1GEA
538
EUR
APOSOR33482-1G
Methyl-4-propionyl-1H-pyrrole-2-carboxylate 97%
Methyl-4-propionyl-1H-pyrrole-2-carboxylate
Formel:
C₉H₁₁NO₃ Molecular Weight: 181.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840964 CAS-Nummer: 1135282-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...