4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-carbaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 4'-(1,3-Dioxolan-2-yl)-[1,1'-biphenyl]-4-carbaldehyde

OR33485-1G
APOSOR33485-1GEA 538 EUR
APOSOR33485-1G
4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-carbaldehyde 95%
4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-carbaldehyde
Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06802589
CAS-Nummer: 893737-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR