2-Methyl-6-(methylamino)-4-pyrimidinol 95%
Lieferant: Apollo Scientific
OR33486-1G
APOSOR33486-1GEA
427
EUR
APOSOR33486-1G
2-Methyl-6-(methylamino)-4-pyrimidinol 95%
2-Methyl-6-(methylamino)-4-pyrimidinol
Formel:
C₆H₉N₃O Molecular Weight: 139,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11840972 CAS-Nummer: 857414-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...