[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol 95%

Lieferant: Apollo Scientific

OR33488-1G
APOSOR33488-1GEA 538 EUR
APOSOR33488-1G
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol 95%
[4'-(1,3-Dioxolan-2-yl)[1,1'-biphenyl]-4-yl]methanol
Formel: C₁₆H₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06802587
CAS-Nummer: 889954-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR