6-(Dimethylamino)-2-methyl-4-pyrimidinol 95%

Lieferant: Apollo Scientific

OR33489-1G
APOSOR33489-1GEA 336 EUR
APOSOR33489-1G
6-(Dimethylamino)-2-methyl-4-pyrimidinol 95%
6-(Dimethylamino)-2-methyl-4-pyrimidinol
Formel: C₇H₁₁N₃O
Molecular Weight: 153.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840974
CAS-Nummer: 83724-17-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR