tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate 95%

Lieferant: Apollo Scientific

OR33490-1G
APOSOR33490-1GEA 874 EUR
APOSOR33490-1G
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate 95%
tert-Butyl-2-(2-chloroacetyl)-1,2-diazepane-1-carboxylate
Formel: C₁₂H₂₁ClN₂O₃
Molecular Weight: 276,77 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840976
CAS-Nummer: 1135283-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR