2,2,2-Trichloro-1-[4-(1-hydroxypropyl)-1H-pyrrol-2-yl]-1-ethanone 95%

Lieferant: Apollo Scientific

OR33491-1G
APOSOR33491-1GEA 538 EUR
APOSOR33491-1G
2,2,2-Trichloro-1-[4-(1-hydroxypropyl)-1H-pyrrol-2-yl]-1-ethanone 95%
2,2,2-Trichloro-1-[4-(1-hydroxypropyl)-1H-pyrrol-2-yl]-1-ethanone
Formel: C₉H₁₀Cl₃NO₂
Molecular Weight: 270,54 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840981
CAS-Nummer: 1135283-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR