5-Chloro-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine 95%

Lieferant: Apollo Scientific

OR33494-1G
APOSOR33494-1GEA 894 EUR
APOSOR33494-1G
5-Chloro-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine 95%
5-Chloro-6-cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-amine
Formel: C₁₀H₁₁ClN₄
Lagertemperatur: Raumtemperatur
MDL: MFCD12025852
CAS-Nummer: 1135283-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR