1-Thia-4-azaspiro[4.4]nonane hydrochloride 95%

Lieferant: Apollo Scientific

OR33499-1G
APOSOR33499-1GEA 339 EUR
APOSOR33499-1G
1-Thia-4-azaspiro[4.4]nonane hydrochloride 95%
1-Thia-4-azaspiro[4.4]nonane hydrochloride
Formel: C₇H₁₄ClNS
Lagertemperatur: Raumtemperatur
MDL: MFCD16140324
CAS-Nummer: 1221792-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR