1,4-Diazaspiro[4.5]decan-2-one 95%
Lieferant: Apollo Scientific
OR33504-1G
APOSOR33504-1GEA
409
EUR
APOSOR33504-1G
1,4-Diazaspiro[4.5]decan-2-one 95%
1,4-Diazaspiro[4.5]decan-2-one
Formel:
C₈H₁₄N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD14584836 CAS-Nummer: 19718-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...