1,2-Dimethyl-1H-1,3-benzodiazol-5-amine 95%

Lieferant: Apollo Scientific

OR33508-1G
APOSOR33508-1GEA 201 EUR
APOSOR33508-1G
1,2-Dimethyl-1H-1,3-benzodiazol-5-amine 95%
1,2-Dimethyl-1H-1,3-benzodiazol-5-amine
Formel: C₉H₁₁N₃
Molecular Weight: 161.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03872210
CAS-Nummer: 3527-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR