3-Methyl-1-benzothiophen-2-amine hydrochloride, tech

Lieferant: Apollo Scientific

OR33524-1G
APOSOR33524-1GEA 1570 EUR
APOSOR33524-1G
3-Methyl-1-benzothiophen-2-amine hydrochloride, tech
3-Methyl-1-benzothiophen-2-amine hydrochloride
Formel: C₉H₉NS·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18904349
CAS-Nummer: 13584-56-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR