3-(1-Benzofuran-2-yl)-3-oxopropanenitrile 95%

Lieferant: Apollo Scientific

OR33529-1G OR33529-5G
APOSOR33529-1GEA 270 EUR
APOSOR33529-1G APOSOR33529-5G
3-(1-Benzofuran-2-yl)-3-oxopropanenitrile 95%
3-(1-Benzofuran-2-yl)-3-oxopropanenitrile
Formel: C₁₁H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00047286
CAS-Nummer: 5149-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR