Methyl-4-(1-hydroxypropyl)-1-methyl-1H-pyrrole-2-carboxylate 95%

Lieferant: Apollo Scientific

OR33531-1G
APOSOR33531-1GEA 874 EUR
APOSOR33531-1G
Methyl-4-(1-hydroxypropyl)-1-methyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-(1-hydroxypropyl)-1-methyl-1H-pyrrole-2-carboxylate
Formel: C₁₀H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12025864
CAS-Nummer: 1135283-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR