(1-Chloro-3,4-dihydro-2-naphthalenyl)methanol 95%
Lieferant: Apollo Scientific
OR33533-1G
APOSOR33533-1GEA
538
EUR
APOSOR33533-1G
(1-Chloro-3,4-dihydro-2-naphthalenyl)methanol 95%
(1-Chloro-3,4-dihydro-2-naphthalenyl)methanol
Formel:
C₁₁H₁₁ClO Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025871 CAS-Nummer: 128104-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...