Methyl-2-(chloromethyl)-5-methyl-1,3-benzoxazole-7-carboxylate 95%

Lieferant: Apollo Scientific

OR33536-1G
APOSOR33536-1GEA 538 EUR
APOSOR33536-1G
Methyl-2-(chloromethyl)-5-methyl-1,3-benzoxazole-7-carboxylate 95%
Methyl-2-(chloromethyl)-5-methyl-1,3-benzoxazole-7-carboxylate
Formel: C₁₁H₁₀ClNO₃
Molecular Weight: 239,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140297
CAS-Nummer: 1221792-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR